PolarSafe® Cooling Transport Boxes, Cole Parmer
Transport your critical samples without worry.
- Maintain specific temperature ranges for extended periods of time for improved sample protection
- Get the temperature protection you need with choice of durable transport bags or boxes
- Modular, color-coded cooling packs allow flexible configurations for different-sized samples
- Ensure sample integrity with optional security sealing ties
Durable polypropylene-insulated walls offer improved insulation for better sample protection. Organize cooling packs to suit your sample or choose from one of our prepared kits. Cooling packs can be added so that no weight is placed on top of your sample (see box images). Boxes are stackable and feature a convenient shoulder strap and integrated handle.
Frequently Bought Together