À propos de cet article
Digital multi-position magnetic stirrer with 6 or 15 positions, that remains cold even after several days of continuous use.
- Bright digital display for accurate speed monitoring
- Exclusive, on-board timer for unattended operation
- Adjustable reverse operation (time-controlled)
- Speed regulation from 80 to 1500 rpm
- SpeedServo™ ensures constant speed even when the viscosity changes
- Synchronized stirring, same speed ensured
- Ergonomic - very low height
- Universal mains voltage/frequency range
Soft start but rapidly reaching set speed, excellent speed control (electronic), even at low speeds, light weight, easy to move. Can thermostat samples using a recirculating water bath. Ideal for microtitration, microbiology and biochemistry.
Stirring system has high-power driving magnet type 'PCM' operated by a mono-phase motor for continuous operation. Distance between stirring position centers for MULTISTIRRER 6 and MULTISTIRRER 15 is 100 and 74 mm.
Auto-reverse from 5 to 900 s ensures enhanced mixing on board timer for unattended operations. Construction material is made up of epoxy painted metal structure; stainless steel top plate. 10 rpm steps speed interval.
Spécifications
- Catalog No:
- 76416-216
- 76416-214
- Model:
- MULTISTIRRER Digital 15
- MULTISTIRRER Digital 6
- Max. stirring capacity H₂O:
- 250 ml
- 400 ml
- Power input:
- 9 W
- 3.6 W
- Weight:
- 2.1 kg (4.6 lbs.)
- 1.75 kg (3.8 lbs.)
- Timer:
- 1 - 900 min
- 1 - 900 min