SlimLine™ Mop Adapters, Micronova
Supplier: Micronova
Certificates
About this item
A lightweight plastic; flat-head mop adapter for use with the SlimLine™, PadMop™, SnapMop™, FlatMop, and PocketMop series.
- Lightweight plastic
- Flathead mop adapter
- Autoclavable
- Compatible with strong disinfectants and common detergents
All frames fit in the SlimLine™ and SlimT™ bucket systems and attach to STA series of plastic handles.
Specifications
- Catalog No:
- 11310-596
- 76214-294
- 11310-600
- 89140-696
- 11310-598
- 10153-678
- 76323-718
- 89140-692
- 89140-694
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing