Accuform Table-Gard Disposable Work Mat, Stitch Pattern
Accuform Table-Gard Disposable Work Mat, Stitch Pattern
Certificates
Accuform Table-Gard Disposable Work Mat, Stitch Pattern
Accuform Table-Gard Disposable Work Mat, Stitch Pattern
3 Options Available Below
Product Details & Documents
Create safe, clean workspaces by applying the adhesive mat to surfaces that are not regularly cleaned.
  • Provides protection from germs, dirt, and chemical residue
  • Compact and lightweight mat fits in workbags for on-the-go working needs
  • Made of a smooth, thin 2-mil plastic film that is waterproof and tear-resistant
  • Can be discarded in waste or recycle bin (recycle code 5)
  • 12.5×19.5" size creates a workspace large enough for laptops and folios
  • BPA, Latex, and PVC-free material without toxic chemicals
  • The adhesive is safe for indirect food contact (complies with FDA 21 CFR 175.105)
The self-adhesive mat is repositionable and lifts easily for disposal when work is complete. The mat is safe for application to countertops, desktops, conference tables, and restaurant tables.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Table-gard disposable work mats, stitch pattern
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $31.24
    $31.24
    Pack of 10
       $31.24
    Supplier Number: LPM127X
  • Table-gard disposable work mats, stitch pattern
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $70.83
    $70.83
    Pack of 25
       $70.83
    Supplier Number: LPM127P
  • Table-gard disposable work mats, stitch pattern
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $118.90
    $118.90
    Pack of 50
       $118.90
    Supplier Number: LPM127L