ContactUsLinkComponent
Double-Sided Polyester Permanent Adhesion Cleanroom Tape with Liner, UltraTape
  89203-824
undefined
Double-Sided Polyester Permanent Adhesion Cleanroom Tape with Liner, UltraTape
  89203-824
 :  1510CL075P3D
  • Width:
    0.75"
  • Color:
    Clear
  • Length:
    36 yards
  • Material:
    UltraClean
  • Cat. no.:
    89203-824
  • Supplier No.:
    1510CL075P3D
  • Adhesive material:
    Acrylic
Particle-free, residue-free tapes.
  • Double-sided permanent adhesive tape
Both sides of the polyester carrier have a transparent red Mylar release liner. Ideal for fastening applications, connecting plastic sheeting, or permanent nameplate mounting.