Detector Lamps for Shimadzu LC Systems for HPLC, Agilent Technologies
CAAG8001-0702
- Description:Agilent deuterium lamp, for Shimadzu SPD-20A/AV and SPD10A/AV/AVvp DAD, 2000 hour, similar to Shimadzu 228-34016-02
- Cat. no.:CAAG8001-0702
- For Use With:Shimadzu SPD-20A/AV and SPD10A/AV/Avvp diode array detectors
- Bulb Life:2000 h
Lamps for selected Shimadzu HPLC systems, compatible with a range of detectors.
- Deuterium lamps provide high output stability and intensity for extended detection capabilities and improved qualification at trace level
- All deuterium lamps have a guaranteed lifetime of 2000 hours
- Tungsten lamps offer coverage in the visible wavelength range for high-sensitivity detection
Agilent lamps for selected Shimadzu HPLC systems are designed and built to be compatible with a wide variety of detectors, including variable wavelength detectors (VWD), multiple wavelength detectors (MWD), diode array detectors (DAD), and photodiode array detectors (PDA). Both deuterium lamps and tungsten lamps are available.
All lamps adhere to the tightest specifications for consistent quality and reproducible performance over the entire lifetime of the lamp.