Bond Elut Certify Plates
Bond Elut certify delivers a cation exchange/mixed-mode SPE approach that is ideal for extraction of drugs from urine and blood, along with many other applications.
- Special mixed-mode sorbent bed
- Broad application range for aqueous extraction
- Bimodal, nonpolar, and strong cation exchange
Bond Elut certify extraction cartridge utilizes a packed bed consisting of a special, nonpolar C8 sorbent and a strong cation exchanger (SCX). Certify is most commonly used to extract basic (cationic) drugs from urine and blood, but it is also very effective for extracting a wide range of compounds from a diverse number of aqueous matrices.
Bond Elut certify delivers a cation exchange/mixed-mode SPE approach that is ideal for extraction of drugs from urine and blood, along with many other applications.
- Special mixed-mode sorbent bed
- Broad application range for aqueous extraction
- Bimodal, nonpolar, and strong cation exchange
Bond Elut certify extraction cartridge utilizes a packed bed consisting of a special, nonpolar C8 sorbent and a strong cation exchanger (SCX). Certify is most commonly used to extract basic (cationic) drugs from urine and blood, but it is also very effective for extracting a wide range of compounds from a diverse number of aqueous matrices.
Product Family Options
- DésignationVolume
- VersaPlate preassembled 96 round-well plate, certify, 50 mg, 40 µm, PE frit (20 µm)1.8 ml-: -
- VersaPlate preassembled 96 round-well plate, certify, 100 mg, 40 µm, PE frit (20 µm)1.8 mlEach$752.00: 7540901C