Extraction Kits, Agilent Technologies | Avantor
Extraction Kits, Agilent Technologies
Extraction Kits, Agilent Technologies
Certificates
Extraction Kits, Agilent Technologies
Extraction Kits, Agilent Technologies
3 Options Available Below
Product Details & Documents
Extraction kits with optional ceramic homogenizers feature salts for buffering of samples.
  • The buffered extraction salts are amenable to more labile pesticides
  • Choose the extraction salt type based on your method of analysis: AOAC or EN
  • Ceramic homogenizers break up salt agglomerates, promoting consistent sample extraction and increasing product recovery during extraction and dispersion
QuEChERS salts and buffers are prepackaged in anhydrous packages. This allows you to add them to the sample after the solvent, based on your method of analysis (AOAC or EN). Extraction kits include: 50 polypropylene centrifuge tubes (50 ml) with caps, MgSO₄, NaCl, or other salts for buffering. Extraction kits can also be purchased with ceramic homogenizers. These provide consistent grinding of the material being homogenized and can minimize variation. Kits featuring ceramic homogenizers are marked with a CH.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Ceramic homogenizers, 2 ml tubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $86.80
    $86.80
    Pack of 100
       $86.80
    Supplier Number: 5982-9311
  • Ceramic homogenizers, 15 ml tubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $87.60
    $87.60
    Pack of 100
       $87.60
    Supplier Number: 5982-9312
  • Ceramic homogenizers, 50 ml tubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $87.60
    $87.60
    Pack of 100
       $87.60
    Supplier Number: 5982-9313