Ultra Inert Press Fit Column Connectors, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Agilent Technologies offers Ultra Inert universal press-fit column connectors.
- Improved packaging and installation instructions
- Ultra Inert deactivation
- Batch certified
In the past it was necessary to use press-fit connectors with specific dimensions to connect columns of those dimensions. Modern press-fit connectors are inlaser-milled in to provide highly reproducible taper angles throughout the length of the press-fit, ensuring an excellent seal. The press-fits are now treated with Agilent Ultra Inert deactivation to provide a robust and inert flow path.
Glass unions can be sealed with polyimide sealing resin which ensures a leak-free and solid connection. This method is recommended if glass press-fit unions are used.
Frequently Bought Together