QuickConnect™ Mop Handles
À propos de cet article
Interchangeable mop handles are available in carbon fiber, anodized aluminum, stainless steel or fiberglass.
- Compatible with all QuickConnect™ frames
- Multiple lengths available
Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments. Invest in one handle and the various interchangeable heads required for specific applications. Handles are available in several lengths and styles.
Telescoping handles are available in lightweight carbon fiber, anodized aluminum, durable stainless steel, or fiberglass. Choose from a variety of lengths. For hard-to-reach ceilings and walls, a carbon fiber handle is available that is lightweight and easy to maneuver. A one-piece stainless-steel handle is also available.
12 Options Available Below
Product Details & Documents
Interchangeable mop handles are available in carbon fiber, anodized aluminum, stainless steel or fiberglass.
- Compatible with all QuickConnect™ frames
- Multiple lengths available
Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments. Invest in one handle and the various interchangeable heads required for specific applications. Handles are available in several lengths and styles.
Telescoping handles are available in lightweight carbon fiber, anodized aluminum, durable stainless steel, or fiberglass. Choose from a variety of lengths. For hard-to-reach ceilings and walls, a carbon fiber handle is available that is lightweight and easy to maneuver. A one-piece stainless-steel handle is also available.
Documents
Product Family Options
Informations sur le produit
- DésignationLongueurAvailabilityPrice
- Supplier Number: 2725E
- QuickConnect™ stainless steel telescopic handle127 - 234 cm (50 - 92")Log in pour la disponibilitéEach$761.10Supplier Number: 2645
- QuickConnect™ anodized aluminum telescopic handle41 - 76 cm (16 - 30")Log in pour la disponibilitéEach$235.64Supplier Number: 2642
- QuickConnect™ anodized aluminum telescopic handle127 - 234 cm (50 - 92")Log in pour la disponibilitéEach$265.74Supplier Number: 2643
- QuickConnect™ stainless steel telescopic handle41 - 76 cm (16 - 30")Log in pour la disponibilitéEach$502.24Supplier Number: 2644
- QuickConnect™ stainless steel handle with hanging loop41 cm (16")Log in pour la disponibilitéEach$50.69Supplier Number: 2686
- QuickConnect™ carbon fiber telescopic handle46 - 81 cm (18 - 32")Log in pour la disponibilitéEach$157.38Supplier Number: CFH32
- QuickConnect™ carbon fiber telescopic handle102 - 178 cm (40 - 70")Log in pour la disponibilitéEach$162.54Supplier Number: CFH70
- QuickConnect™ carbon fiber telescopic handle160 - 290 cm (63 - 114")Log in pour la disponibilitéEach$236.50Supplier Number: CFH114
- QuickConnect™ carbon fiber telescopic handle168 - 568 cm (66 - 224")Log in pour la disponibilitéEach$447.20Supplier Number: CFH224
- QuickConnect™ carbon fiber telescopic handle175 - 848 cm (69 - 334")Log in pour la disponibilitéEach$627.80Supplier Number: CFH334
- Supplier Number: CFH18