Capillary Cassettes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Quick-change capillary cassettes that enable capillary exchange in less than a minute.
- The quick-change cassette allows capillary exchange in less than a minute
- The cassette and interfaces accept all commercially available capillaries with a 365 µm OD
Agilent provides quick-change capillary cassettes that enable capillary exchange in less than a minute. The color code of the alignment interface must match with the color code of the capillary's built-in alignment stopper.
Frequently Bought Together