ADM Flow Meter, Agilent Technologies | Avantor
ADM Flow Meter, Agilent Technologies
ADM Flow Meter, Agilent Technologies
Certificates
ADM Flow Meter, Agilent Technologies
ADM Flow Meter, Agilent Technologies
2 Options Available Below
Product Details & Documents
Battery-powered and portable flow meter for measuring gas streams with changing composition.
  • Valuable tool for troubleshooting detector problems
  • Recalibrate annually by replacing cartridge with a NIST-certified calibration cartridge
  • Measures gas flow volumetrically, eliminating the need to select a gas type
  • USB connects to web interface for updates and monitoring
  • Easy to view OLED screen
  • New composite materials construction
The Agilent ADM flow meter is ideal for measuring gas streams with composite gas composition. This digital flow meter is easy to maintain by simply replacing the cartridge once a year to keep the gas flow meter compliant. There is no need to ship your meter back for recalibration, simply order and replace the cartridge yourself. Each flow meter comes with a calibrated cartridge.
 : Each flow meter comes with a calibrated cartridge.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • ADM flow meter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 450,00 $
    1 450,00 $
    Each
       1 450,00 $
    Supplier Number: G6691A
  • ADM flow meter, made to order
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 480,00 $
    1 480,00 $
    Each
       1 480,00 $
    Supplier Number: G6691E
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • FID adapter, for flow meter
    FID adapter, for flow meter
    CA5190-9576
    Log in pour la disponibilité
    Each - $93.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement cartridge, for ADM flow meter
    Replacement cartridge, for ADM flow meter
    CAG6692A
    Log in pour la disponibilité
    Each - $339.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Split vent adapter, for flow meter
    Split vent adapter, for flow meter
    CA5190-9580
    Log in pour la disponibilité
    Each - $93.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Carrying case, for ADM flow meter
    Carrying case, for ADM flow meter
    CAG6691-40500
    Log in pour la disponibilité
    Each - $99.30
Informations sur le produit
  • Catalog #
    Availability
    Price
  • TCD adapter, for flow meter
    TCD adapter, for flow meter
    CAAG5190-9578
    Log in pour la disponibilité
    Each - $93.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement cartridge, for ADM flow meter, made to order
    Replacement cartridge, for ADM flow meter, made to order
    CAAGG6692E
    Log in pour la disponibilité
    Each - $392.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • ECD adapter, for flow meter
    ECD adapter, for flow meter
    CAAG5190-9579
    Log in pour la disponibilité
    Each - $93.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • NPD adapter, for flow meter
    NPD adapter, for flow meter
    CAAG5190-9577
    Log in pour la disponibilité
    Each - $93.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement ADM cartridge, with assessment
    Replacement ADM cartridge, with assessment
    CAAGG6692X
    Log in pour la disponibilité
    Each - $508.00