Polypropylene Vials, Agilent Technologies
Polypropylene Vials, Agilent Technologies
Certificates
Polypropylene Vials, Agilent Technologies
Polypropylene Vials, Agilent Technologies
5 Options Available Below
Product Details & Documents
These vials are appropriate for pH sensitive samples, sodium, and heavy metal analyses.
  • Manufactured from virgin PP
  • Chemically resistant
Wide opening glass/plastic vials have a glass flanged insert molded to the inside, so sample only comes in contact with high quality glass insert and septum.
 : Meets requirements of 21 CFR 177.1520.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Diameter ×H
    Matière
    Color
    Avec bouchon
    Taille du bouchon
    Availability
    Price
  • Crimp/snap top vials
    1.00 ml
    12×32 mm
    PP
    Clear
    Non
    11 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $33.70
    $33.70
    Pack of 100
       $33.70
    Supplier Number: 5182-0567
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76508-116
  • Crimp/snap top vials
    0.25 ml
    12×32 mm
    PP
    Clear
    Non
    11 mm
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    CA5190-3155
  • Crimp/snap top vials with glass insert
    0.25 ml
    12×32 mm
    PP
    Clear
    Non
    11 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $333.00
    $333.00
    Pack of 100
       $333.00
    Supplier Number: 9301-0977
    Spécifications:
    More
    Specifications
    Cat. No.
    CA9301-0977
  • Screw top vials with fixed glass insert
    0.25 ml
    12×32 mm
    PP
    Non
    12 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $344.00
    $344.00
    Pack of 100
       $344.00
    Supplier Number: 5188-5390
    Spécifications:
    More
    Specifications
    Cat. No.
    CA5188-5390
  • Screw top vials, certified
    0.25 ml
    12×32 mm
    PP
    Non
    12 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $50.70
    $50.70
    Pack of 100
       $50.70
    Supplier Number: 5190-2242
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG5190-2242
  • Screw top vials, certified
    0.25 ml
    12×32 mm
    PP
    Non
    12 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1,000
    Pack of 1,000 - $439.00
    $439.00
    Pack of 1,000
       $439.00
    Supplier Number: 5190-2243
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG5190-2243