Kimtech™ Disposable N95 Pouch Respirator, Kimberly-Clark Professional
Catalogue # 76420-910
Fournisseur: Kimberly-Clark
Spécifications
- Color:White with blue headband
- Description:N95 respirator
- Size:Universal
- Cat. no.:76420-910
Spécifications
À propos de cet article
The Kimtech™ N95 pouch respirator is designed to help guard against the risks posed by airborne particles and contaminants.
- NIOSH-approved as N95 particulate filter respirator (TC84A-9042)
- Pouch design with a large breathing chamber for added comfort
- Consistent seal to ensure proper fit
- Comfort fit headband
- Bendable nose piece
- Made in the USA, TAA-compliant
This respirator is NIOSH-certified as an N95 particulate filter respirator, demonstrating that it provides a minimum 95% filtration efficiency of 0.3 micron particles (though not oil resistant).
The pouch-style respirator offers the wearer a large breathing chamber to maximize comfort. In addition, the N95 comes with a comfort fit head band and a bendable nosepiece to customize fit and ensure a consistent seal.
Certifications: NIOSH-approved as N95 particulate filter respirator (TC84A-9042).
Caution: This product is not for Medical Use and has not been tested against COVID-19 so cannot be recommended for efficacy or use.