Ward's® Individual Dissection Specimens | Avantor
Ward's® Individual Dissection Specimens
Supplier:   Avantor
Ward's® Individual Dissection Specimens
Supplier:  Avantor
Certificates
Ward's® Individual Dissection Specimens
Ward's® Individual Dissection Specimens
1 Options Available Below
Product Details & Documents
Ideal for distance learning or homeschooling, these individually wrapped specimens have low odor with high quality preservation.
  • Safest specimens
  • Low formalin odor
  • Best classroom experience
Ward's® PURE preserved specimens offer the best classroom experience because PURE specimens are safe. Students stay focused on learning and not on smells. Experience quality with true-to-life preservation, combined with artfully illustrated dissection guides, free with every purchase, to maximize value, lesson time and student engagement.
We guarantee you’ve never experienced dissection like this before. Go Pure.
Packaging: Individually packed.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Injection Type
    Packaging
    Availability
    Price
  • Lubber grasshopper (Romalea microptera)
    3"
    Uninjected
    Jar of 1
    Discontinued
  • Large earthworm (Lumbricus terrestris)
    9"+
    Uninjected
    Jar of 1
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $13.99
    $13.99
    Each
       $13.99
  • Freshwater perch (Perca flavescens)
    5 - 7"
    Uninjected
    Vacuum pack of 1
    Discontinued
  • Long neck clam (Mya arenaria)
    1.5 - 2"
    Uninjected
    Jar of 1
    Discontinued