PIG® WorkWipes® Series 30 Wipers, NEW PIG
Catalog # 89073-939
Supplier: New Pig
Specifications
- Description:WorkWipes® series 30 wipers
- Dimensions:11.3×13"
- Size:30×960
- Cat. no.:89073-939
- Form:Wipers
- Supplier No.:WIP630
Specifications
About this item
This lighter version of WorkWipes series 40 aces general housekeeping. Plus cloth-like softness for wiping hands and face.
- Good for lighter-duty jobs like wiping up small spills or cleaning hand tools
- Clothlike construction is soft enough for wiping hands and face
- Made with binder forced deep into soft cellulose fibers for extra durability and strength
- Wipers are ¹/₄ folded and poly wrapped for easy handling and loading
Great for manufacturing facilities and general housekeeping applications.