ContactUsLinkComponent
PIG® Retract-a-Clamp® Insulated Bonding/Grounding Wires, New Pig
PIG® Retract-a-Clamp® Insulated Bonding/Grounding Wires, New Pig
  75839-192
 :  New Pig
undefined
PIG® Retract-a-Clamp® Insulated Bonding/Grounding Wires, New Pig
  75839-192
 :  New Pig
 :  DRM416
  • Description:
    Retract-a-Clamp® insulated bonding or grounding wires, hand clamp grounding end, hand clamp bonding end, 10'
  • Cat. no.:
    75839-192
  • Supplier No.:
    DRM416
Wire creates safe grounding connection while staying coiled to avoid creating a tripping hazard.
  • Bonding and grounding assembly controls static electricity to prevent sparking while transferring or storing flammable liquids; FM approved and NFPA 77 compliant
  • Vinyl coating protects wire from moisture and corrosion for longer life and to allow outdoor use
Wire creates a solid grounding connection while staying coiled so as not to be a tripping hazard.