Ionizer with Electrode, Sartorius
Ionizer with U-shaped electrode, 115 V, 0.935 kg
Catalog # CA76413-012
Supplier: Sartorius
Fournisseur #: YIB02-115V
Jump to:
Product Details & Documents
Ionizer with U-shaped electrode, used with laboratory weighing balances.
Delivery information: Includes power supply (115 V)
Spécifications
- Désignation:Ionizer with U-shaped electrode
- Electrical:115 V
- Weight:0.935 kg