Plastic Chain S-Hooks, Accuform®
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
À propos de cet article
Plastic chain S-hooks are for use with plastic chain to quickly link and unlink separate lengths of plastic chain.
- Lightweight, yet remarkably strong
- Polyethylene plastic will not rust or corrode
- Each link is 2×1×¹/₄"
Frequently Bought Together