Bucket of Plastic Chains, Accuform®
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
À propos de cet article
Bucket of plastic chains allows you to set up a barrier quickly and economically when used with stanchion posts (sold separately).
- Lightweight, yet remarkably strong
- Polyethylene plastic will not rust or corrode
- Good for limited outdoor use
- 5-gallon bucket contains 100 ft. of chain for easy storage
- Each link is 2×1×¹/₄"
Frequently Bought Together