Standard, stainless steel spring tethers without luers or tubing. Used with SAI Standard Harness (SAH) and skin buttons.
- Durable, reusable, and affordable
- Catheter fit will depend on wall thickness, material, and elasticity
- Sizing kit (not included) will properly fit harness to catheter
- Custom sizes are also available
Light weight tethers protect the catheter line for the duration of an infusion study. Pair with a swivel and harness or skin button, and choose the appropriate length for the study. Additional options are available for mice and other infusion setups.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.