About this item
SAI extension lines are ideal for connecting between pump and swivel or pump and catheter in animal infusion studies. Made with non-DEHP PVC tubing, extension lines include one male and one female luer adaptor, and are available in a variety of standard lengths.
- Lines feature one male luer end and one female luer end
- ETO sterilized
- High flow and micro flow options available
- Custom sizes are also available
SAI's luered extension lines are compatible with any standard luer fitting, providing a simple, quick, and secure luer connection. These luered fittings offer unlimited flexibility for research and infusion needs. Designed for animal research use only.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.