About this item
Clean and Sweep Store-Boards™ Kit keeps tools organized.
- Promote the concept, a place for everything and everything in its place
- At a glance, see that tools and supplies are in their designated storage location
- Tools included
- Material options: Accu-Shield™, Ultra Aluma-Lite™ or Max-Duty Aluminum
- Board measurement: 172.72H×76.2W cm
Accu-Shield™ - Highest performance material, is a two-ply, polycarbonate plastic. Clear, non-glare front provides heavy duty protection against chemicals, corrosion and abrasions. Subsurface, UV-printed image, encapsulated on the back by poly laminate, will continue to look good for years while maintaining superior strength. Service temperature: –60 to 260 °F.
Ultra Aluma-Lite™ - Composite plastic bonded with two layers of aluminum making it light-weight, 40% lighter than solid aluminum, but extremely durable. Corrosion-, impact-, and UV-resistant making it an ideal choice for indoor or outdoor use. Service temperature: –40 to 170 °F.
Max-Duty Aluminum - Provides the strength of metal yet never rusts. Finishing includes UV protective coating to prevent fading, chemical damage, and abrasions. Service temperature: -40° to 250 °F.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.