Tough-Mark™ Heavy Duty Floor Marking Strips, Accuform
Supplier: ACCUFORM MANUFACTURING, INC
Certificates
About this item
Tough-Mark™ heavy duty floor marking strips mark specific areas in the workplace to keep clear of obstructions and debris, designate storage locations, and create aisles, boundaries, and pathways for safe accessibility.
- Easy to apply and bonds to a variety of clean, sealed, and non-coated surfaces
- Made with 55 mil polyethylene with 9.4 mil thick polypropylene adhesive backing
- Removes easily while staying in one piece
- 48" long strips allow you to keep lines straight
- Can be trimmed to needed length or to create corners
The beveled edges of the strip reduce tears and lifting commonly caused by pallet dragging.
Frequently Bought Together