Plastic Tube Brushes, Micronova
Supplier: Micronova
Certificates
About this item
These durable, 100% polypropylene brushes clean finely polished stainless steel parts without scratching.
- 100% polypropylene bristles and handle
- Thin core and handles are flexible enough to bend through fittings
- Available in a range of bristle diameters and handle lengths
Brushes feature narrow center cores that are flexible enough to pass through elbow tank fittings.
Certifications: FDA compliant materials.
Frequently Bought Together