Bertin Instruments User Maintenance Kit for Precellys Evolution, Bertin Corp
Precellys Evolution maintenance kit
Catalog # 76415-486
Supplier: BERTIN CORP
Fournisseur #: S000808-PEVO0-A.0
Bertin Instruments User Maintenance Kit for Precellys Evolution, Bertin Corp
Catalog # 76415-486
Supplier: BERTIN CORP
Jump to:
Product Details & Documents
Precellys Evolution maintenance kit contains all the external pieces to replace each year of use allowing to give the maximum life time to the equipment.
- Allows you to enhance the life time of your Precellys Evolution equipment
For Precellys Evolution, Contains: 2× vacuum joints, 1× containment seal, 1× anti-rotation tubing kit.
Spécifications
- Désignation:Precellys Evolution maintenance kit