Corning® Matrigel® Matrix-3D Plates, Corning
Fournisseur: Corning
Certificats
À propos de cet article
A Matrigel® pre-coated plate which can be used for drug discovery applications with 3D cell cultures such as organoids, spheroids or tumoroids.
- Available in 384 and 96-well ready-to-use formats
- Easy and convenient as Matrigel® is pre-dispensed in each well
- Formats provide intra-plate consistency
- Allows for sandwich/overlay and embedded application assays
The Corning® Matrigel® matrix-3D plate is a high-throughput pre-coated format that contains Matrigel® and can be used for drug discovery applications with 3D cell cultures such as organoids, spheroids or tumoroids. Matrigel® Matrix is extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors.
Each lot is verified for intraplate consistency. Plates are tested and found negative for the presence of bacteria and fungi.
Spécifications
- Catalog No:
- 76419-680
- 76419-678
- 76419-684
- 76419-682
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 - Bedford (reg. no. 14.4942)
- ISO 14001 - Bedford (reg. no. 14.4942)
- ISO 14001 - Bedford (reg. no. 14.4942)