VWR® Screw Microtubes with Flat Caps
VWR® Screw Microtubes with Flat Caps
VWR® Screw Microtubes with Flat Caps
VWR® Screw Microtubes with Flat Caps
VWR® Screw Microtubes with Flat Caps
VWR® screw cap microtubes are the perfect environment to securely store your important samples and reagents.
  • All tubes are manufactured from ultra-pure PP
  • High-clarity tubes provide excellent visibility to the sample
  • O-ringed flat caps feature a ³/₄ turn to open or close
  • Wide temperature range to meet a wide variety of applications: −196 to +121 °C, but not suitable for immersion in liquid nitrogen
  • Sterile assemblies or handy, non sterile combo packs are available
Screw cap microtubes are available in the three most commonly volumes and available with conical or free-standing/skirted bottoms.
Certifications: Certified free of detectable DNase, RNase, DNA, PCR inhibitors. Sterile items are also free of pyrogens. The PP used in tubes meets FDA 21 CFR 177.1520 & USP Class VI standards.
Documents
Product Family Options
Screw cap microtubes without caps
  • Désignation
    Color
    Volume
    Cap Color
    Format
    Cap Type
    Graduation
    Maximum RCF
    Packaging
    Sterility
  • Microtubes without ribbing
    Clear
    0.5 ml
    Conical bottom
    No
    20000×g
    Non sterile
    Pack of 500
    Pack of 500 - $114.08
    $114.08
    Case of 5000
    Case of 5000 - $1,140.80
    $1,140.80
    Pack of 500
       $114.08
    Case of 5000
       $1,140.80
  • Microtubes without ribbing
    Clear
    0.5 ml
    Freestanding
    No
    20000×g
    Non sterile
    Pack of 500
    Pack of 500 - $114.08
    $114.08
    Case of 5000
    Case of 5000 - $1,140.80
    $1,140.80
    Pack of 500
       $114.08
    Case of 5000
       $1,140.80
  • Microtubes without ribbing
    Clear
    1.5 ml
    Conical bottom
    Yes
    20000×g
    Non sterile
    Pack of 500
    Pack of 500 - $127.65
    $127.65
    Case of 5000
    Case of 5000 - $1,276.48
    $1,276.48
    Pack of 500
       $127.65
    Case of 5000
       $1,276.48
  • Microtubes without ribbing
    Clear
    1.5 ml
    Freestanding
    No
    20000×g
    Non sterile
    Pack of 500
    Pack of 500 - $127.65
    $127.65
    Case of 5000
    Case of 5000 - $1,276.48
    $1,276.48
    Pack of 500
       $127.65
    Case of 5000
       $1,276.48
  • Microtubes without ribbing
    Clear
    1.5 ml
    Conical bottom
    No
    20000×g
    Non sterile
    Pack of 500
    Pack of 500 - $129.58
    $129.58
    Case of 5000
    Case of 5000 - $1,295.84
    $1,295.84
    Pack of 500
       $129.58
    Case of 5000
       $1,295.84
  • Microtubes without ribbing
    Clear
    2.0 ml
    Conical bottom
    No
    20000×g
    Non sterile
    Pack of 500
    Pack of 500 - $142.96
    $142.96
    Case of 5000
    Case of 5000 - $1,429.60
    $1,429.60
    Pack of 500
       $142.96
    Case of 5000
       $1,429.60
  • Microtubes without ribbing
    Clear
    2.0 ml
    Freestanding
    No
    20000×g
    Non sterile
    Pack of 500
    Pack of 500 - $140.85
    $140.85
    Case of 5000
    Case of 5000 - $1,408.48
    $1,408.48
    Pack of 500
       $140.85
    Case of 5000
       $1,408.48
  • Microtubes without ribbing
    Clear
    2.0 ml
    Conical bottom
    Yes
    20000×g
    Non sterile
    Pack of 500
    Pack of 500 - $114.31
    $114.31
    Case of 5000
    Case of 5000 - $1,143.06
    $1,143.06
    Pack of 500
       $114.31
    Case of 5000
       $1,143.06
  • Microtubes without ribbing
    Clear
    2.0 ml
    Freestanding
    Yes
    20000×g
    Non sterile
    Pack of 500
    Pack of 500 - $127.18
    $127.18
    Case of 5000
    Case of 5000 - $1,271.84
    $1,271.84
    Pack of 500
       $127.18
    Case of 5000
       $1,271.84
  • Microtubes with ribbing
    Clear
    0.5 ml
    Freestanding
    No
    20000×g
    Non sterile
    Pack of 500
    Pack of 500 - $117.62
    $117.62
    Case of 5000
    Case of 5000 - $1,176.24
    $1,176.24
    Pack of 500
       $117.62
    Case of 5000
       $1,176.24
  • Microtubes with ribbing
    Clear
    1.5 ml
    Conical bottom
    No
    20000×g
    Non sterile
    Pack of 500
    Pack of 500 - $127.65
    $127.65
    Case of 5000
    Case of 5000 - $1,276.48
    $1,276.48
    Pack of 500
       $127.65
    Case of 5000
       $1,276.48
  • Microtubes with ribbing
    Clear
    1.5 ml
    Freestanding
    No
    20000×g
    Non sterile
    Pack of 500
    Pack of 500 - $127.65
    $127.65
    Case of 5000
    Case of 5000 - $1,276.48
    $1,276.48
    Pack of 500
       $127.65
    Case of 5000
       $1,276.48
  • Microtubes with ribbing
    Clear
    2.0 ml
    Conical bottom
    No
    20000×g
    Non sterile
    Pack of 500
    Pack of 500 - $129.91
    $129.91
    Case of 5000
    Case of 5000 - $1,299.12
    $1,299.12
    Pack of 500
       $129.91
    Case of 5000
       $1,299.12
  • Microtubes with ribbing
    Clear
    2.0 ml
    Freestanding
    No
    20000×g
    Non sterile
    Pack of 500
    Pack of 500 - $129.91
    $129.91
    Case of 5000
    Case of 5000 - $1,299.12
    $1,299.12
    Pack of 500
       $129.91
    Case of 5000
       $1,299.12
Screw cap microtubes - Sterile assemblies with flat caps
  • Désignation
    Color
    Volume
    Cap Color
    Format
    Cap Type
    Graduation
    Maximum RCF
    Packaging
    Sterility
  • Clear
    0.5ml
    Clear
    Conical base
    Flat cap
    No
    20000×g
    Sterile
    Pack of 500
    Pack of 500 - $156.13
    $156.13
    Case of 5000
    Case of 5000 - $1,561.28
    $1,561.28
    Pack of 500
       $156.13
    Case of 5000
       $1,561.28
    New Product
    New Product
  • Clear
    1.5 ml
    Clear
    Freestanding
    Flat cap
    No
    20000×g
    Combo pack
    Non sterile
    Pack of 500
    Pack of 500 - $98.86
    $98.86
    Case of 5000
    Case of 5000 - $988.64
    $988.64
    Pack of 500
       $98.86
    Case of 5000
       $988.64
    New Product
    New Product
  • Clear
    0.5 ml
    Clear
    Freestanding
    Flat cap
    No
    20000×g
    Sterile
    Pack of 500
    Pack of 500 - $453.22
    $453.22
    Case of 5000
    Case of 5000 - $4,532.24
    $4,532.24
    Pack of 500
       $453.22
    Case of 5000
       $4,532.24
  • Clear
    0.5 ml
    Clear
    Freestanding
    Flat cap
    No
    20000×g
    Sterile
    Pack of 500
    Pack of 500 - $453.22
    $453.22
    Case of 5000
    Case of 5000 - $4,532.24
    $4,532.24
    Pack of 500
       $453.22
    Case of 5000
       $4,532.24
  • Clear
    1.5 ml
    Clear
    Conical bottom
    Flat cap
    Yes
    20000×g
    Sterile
    Pack of 500
    Pack of 500 - $432.33
    $432.33
    Case of 5000
    Case of 5000 - $4,323.30
    $4,323.30
    Pack of 500
       $432.33
    Case of 5000
       $4,323.30
  • Clear
    1.5 ml
    Clear
    Freestanding
    Flat cap
    No
    20000×g
    Sterile
    Pack of 500
    Pack of 500 - $442.90
    $442.90
    Case of 5000
    Case of 5000 - $4,429.04
    $4,429.04
    Pack of 500
       $442.90
    Case of 5000
       $4,429.04
  • Clear
    2.0 ml
    Clear
    Conical bottom
    Flat cap
    Yes
    20000×g
    Sterile
    Pack of 500
    Pack of 500 - $435.67
    $435.67
    Case of 5000
    Case of 5000 - $4,356.74
    $4,356.74
    Pack of 500
       $435.67
    Case of 5000
       $4,356.74
  • Clear
    2.0 ml
    Clear
    Freestanding
    Flat cap
    Yes
    20000×g
    Sterile
    Pack of 500
    Pack of 500 - $439.91
    $439.91
    Case of 5000
    Case of 5000 - $4,399.14
    $4,399.14
    Pack of 500
       $439.91
    Case of 5000
       $4,399.14
Screw cap microtubes with flat caps in separate resealable bags
  • Désignation
    Color
    Volume
    Cap Color
    Format
    Cap Type
    Graduation
    Maximum RCF
    Packaging
    Sterility
  • Clear
    2.0 ml
    Clear
    Conical base
    Flat cap
    Yes
    20000×g
    Combo pack
    Non sterile
    Pack of 500
    Pack of 500 - $95.41
    $95.41
    Case of 5000
    Case of 5000 - $954.08
    $954.08
    Pack of 500
       $95.41
    Case of 5000
       $954.08
    New Product
    New Product
  • Clear
    2.0 ml
    Clear
    Freestanding
    Flat cap
    Yes
    20000×g
    Combo pack
    Non sterile
    Pack of 500
    Pack of 500 - $95.41
    $95.41
    Case of 5000
    Case of 5000 - $954.08
    $954.08
    Pack of 500
       $95.41
    Case of 5000
       $954.08
    New Product
    New Product
  • Clear
    0.5 ml
    Clear
    Conical base
    Flat cap
    No
    20000×g
    Combo pack
    Non sterile
    Pack of 500
    Pack of 500 - $98.86
    $98.86
    Case of 5000
    Case of 5000 - $988.64
    $988.64
    Pack of 500
       $98.86
    Case of 5000
       $988.64
    New Product
    New Product
  • Clear
    0.5 ml
    Clear
    Freestanding
    Flat cap
    No
    20000×g
    Combo pack
    Non sterile
    Pack of 500
    Pack of 500 - $103.87
    $103.87
    Case of 5000
    Case of 5000 - $1,038.72
    $1,038.72
    Pack of 500
       $103.87
    Case of 5000
       $1,038.72
    New Product
    New Product
  • Clear
    1.5 ml
    Clear
    Conical base
    Flat cap
    Yes
    20000×g
    Combo pack
    Non sterile
    Pack of 500
    Pack of 500 - $98.86
    $98.86
    Case of 5000
    Case of 5000 - $988.64
    $988.64
    Pack of 500
       $98.86
    Case of 5000
       $988.64
    New Product
    New Product
Screw cap microtubes with flat caps in separate resealable bags
  •  
    Sterility
    Cap Color
  • Non sterile
    Opaque blue
    Pack of 500
    Pack of 500 - $235.79
    $235.79
    Case of 5000
    Case of 5000 - $235.79
    $235.79
    Pack of 500
       $235.79
    Case of 5000
       $235.79
  •  
    Sterility
    Cap Color
  • Non sterile
    Clear
    Pack of 500
    Pack of 500 - $238.40
    $238.40
    Case of 5000
    Case of 5000 - $238.40
    $238.40
    Pack of 500
       $238.40
    Case of 5000
       $238.40
  •  
    Sterility
    Cap Color
  • Non sterile
    Opaque orange
    Pack of 500
    Pack of 500 - $235.79
    $235.79
    Case of 5000
    Case of 5000 - $235.79
    $235.79
    Pack of 500
       $235.79
    Case of 5000
       $235.79
  •  
    Sterility
    Cap Color
  • Non sterile
    Opaque red
    Pack of 500
    Pack of 500 - $235.79
    $235.79
    Case of 5000
    Case of 5000 - $235.79
    $235.79
    Pack of 500
       $235.79
    Case of 5000
       $235.79
  •  
    Sterility
    Cap Color
  • Non sterile
    Opaque green
    Pack of 500
    Pack of 500 - $235.79
    $235.79
    Case of 5000
    Case of 5000 - $235.79
    $235.79
    Pack of 500
       $235.79
    Case of 5000
       $235.79
  •  
    Sterility
    Cap Color
  • Non sterile
    Opaque yellow
    Pack of 500
    Pack of 500 - $235.79
    $235.79
    Case of 5000
    Case of 5000 - $235.79
    $235.79
    Pack of 500
       $235.79
    Case of 5000
       $235.79
  •  
    Sterility
    Cap Color
  • Non sterile
    Opaque violet
    Pack of 500
    Pack of 500 - $235.79
    $235.79
    Case of 5000
    Case of 5000 - $235.79
    $235.79
    Pack of 500
       $235.79
    Case of 5000
       $235.79
  •  
    Sterility
    Cap Color
  • Non sterile
    Assorted
    Pack of 500
    Pack of 500 - $235.79
    $235.79
    Case of 5000
    Case of 5000 - $235.79
    $235.79
    Pack of 500
       $235.79
    Case of 5000
       $235.79