Blue Phantom® Breast Elastography Ultrasound Trainer
Fournisseur: ELEVATE HEALTHCARE, INC. SE
Certificats
À propos de cet article
A broad range of elastic masses.
- Hypoechoic, isoechoic, as well as echogenic (hyperechoic) echotextures
- Masses range from 6 to 11 mm
- Self-healing tissue
- Varying sized masses
- Feels and biopsies like human tissue
- Masses in central breast tissue and the tail of spence
- Use with any ultrasound system
Blue Phantom® breast elastography ultrasound training model allows users to develop and practice the skills necessary to gain proficiency in using ultrasound for elastography imaging and guided surgical procedures. Containing a broad range of elastic masses, our models allow users to gain experience using stiff, soft, as well as isoelastic lesions - all in the same model. These masses have echotextures that are hypoechoic, isoechoic, as well as echogenic (hyperechoic). The ultra-durable models do not require special storage and will not dehydrate or decompose over time which saves money.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Frequently Bought Together