HyClone™ Characterized Fetal Bovine Serum, CA Origin, HyClone products (Cytiva)
: Cytiva
A cost-efficient alternative to defined FBS, this high-quality Canadian serum is well-characterized.
- Characterized: assayed for a variety of critical components and biochemical characteristics
- Canadian origin: sourced in Canada and processed in the US with complete traceability
- Endotoxin-tested: certificate of analysis confirms ≤25 EU/ml endotoxin
- Tripled-filtered: serial 100 nm pore size-rated filterings for reliable microbial depletion
γ-irradiated and heat-inactivated FBS options also available.
: Broad virus panel testing according toin accordance with 9 CFR 113.53
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- CA76324-898
- CA76324-900
- CA76324-932
- Origin:
- Animal
- Animal
- Animal
- Endotoxin:
- ≤25 EU/ml
- ≤25 EU/ml
- ≤25 EU/ml
- Hemoglobin:
- ≤25 mg/dl
- ≤25 mg/dl
- ≤25 mg/dl
- Storage temperature:
- Less than –10 °C
- Less than –10 °C
- Less than –10 °C
Frequently Bought Together