HyClone™ Defined Fetal Bovine Serum (FBS), US Origin, HyClone products (Cytiva)
Fournisseur: Cytiva
Certificats
À propos de cet article
Highest quality FBS, HyClone US-origin defined FBS is 40 nm-filtered, virus-tested to 9 CFR standard, and supplied with an extensive biochemical profile.
- Well-defined: assay results for over 50 components, with additional testing available
- US origin: sourced and processed in the United States with complete traceability to origin
- Ultra-low endotoxin: ≤10 EU/ml endotoxin for reduced research and production risk
- Sterile-filtered: serial 40 nm filter process removes up to 8 logs viral challenge
- No mycoplasma is present
γ-irradiated and heat-inactivated FBS options also available.
Certifications: Broad virus panel testing in accordance with 9 CFR 113.53; Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- CA76324-860
- CA76324-920
- CA76324-858
- CA76324-918
- Origin:
- Animal
- Animal
- Animal
- Animal
- Endotoxin:
- ≤10 EU/ml
- ≤10 EU/ml
- ≤10 EU/ml
- ≤10 EU/ml
- Hemoglobin:
- ≤10 mg/dl
- ≤10 mg/dl
- ≤10 mg/dl
- ≤10 mg/dl
- Storage temperature:
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
- Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.