Construction paper | Avantor
Construction paper
Assorted colors, Package of 50, 9×12"
Catalog # 470057-784
Supplier:   FUN EXPRESS
Fournisseur #:  3L-73/64775
Construction paper
Catalog # 470057-784
Supplier:  FUN EXPRESS
Construction paper
Assorted colors, Package of 50, 9×12"
Price
$6.30
Log in pour la disponibilité
Pack of 50 Case of 2500
Product Details & Documents
Spécifications
  • Désignation:
    Assorted colors
  • Dimensions:
    9×12"
  • Packaging:
    Package of 50