ContactUsLinkComponent
Printed Graduations Beakers with Handle, PMP, United Scientific Supplies
Catalogue # 76414-738
Fournisseur #: P50908
undefined
Printed Graduations Beakers with Handle, PMP, United Scientific Supplies
Catalogue # 76414-738
Supplier Number:  P50907
Fournisseur #: P50908
S'identifier pour la disponibilité
Select an Option
10000 ml
Select an Option
330 mm
Select an Option
76414-738
À propos de cet article
Includes rectangular handle for easy carrying and pouring.
  • Easy to read silk-screened graduations
  • Autoclavable
Polymethylpentene beakers with pouring spouts are ideal for general laboratory use.
Frequently Bought Together