Static Pen, Sartorius
Stat-Pen
Catalog # CA76413-056
Supplier: Sartorius
Supplier #: YSTP01
Jump to:
Product Details & Documents
This pen unit is used to neutralize static electricity on samples or objects to be weighed down to a level of 30V.
The pen features a micro switch to activate the function, main 100 to 230V with 0.1W, 50/60 Hz.
Spécifications
- Désignation:Stat-Pen