$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
Furniture
Storage Cabinets
Accessories for Diversified Tote Tray and Shelving Storage Cabinet
Accessories for Diversified Tote Tray and Shelving Storage Cabinet
Accessories for Diversified Tote Tray and Shelving Storage Cabinet
Catalog # 470018-184
Supplier:
Diversified Woodcrafts
Expand image
Accessories for Diversified Tote Tray and Shelving Storage Cabinet
Catalog # 470018-184
Supplier:
Diversified Woodcrafts
Supplier Number:
100135
Specifications
Description:
Tote tray replacement, 19D×3½H×14¼"W
Material:
Plastic
Cat. no.:
470018-184
Supplier No.:
100135
Specifications
About this item
Product Details
Product Details