VWR® E-Series Balances with Calibration Certificate
Fournisseur: VWR International
Certificats
À propos de cet article
The VWR® E-Series/C balances with calibration certificate are the answer for an organizations regulatory and compliance needs.
- Complete calibration data reported
- Backlit, LCD screen
- External calibration
- RS232 communication interface
- Five year warranty
The portable balances include a Certificate of Calibration showing the full testing data from an ISO/IEC 17025:2017 accredited Calibration and Testing Laboratory utilizing NIST-traceable weights. These economical and versatile balances offer a variety of standard features, such as multiple weighing units, sturdy construction, simple operation, and quick stabilization time.
The E-series has external calibration and requires purchase of mass standards to perform calibration procedure.
Certifications: ISO/IEC 17025:2017 Accredited Calibration and Testing Laboratory issues Calibration Certificate; CE marked.
Ordering information: The VWR-10001E and VWR-20001E models feature an internal rechargeable battery. All other units utilize 6×AA batteries. All units supplied with switching AC adapter, weighing pan, and user manual. Includes five-year warranty. Accessories not included.
Spécifications
- Catalog No:
- 76378-452
- 76378-454
- 76378-350
- 76378-456
- 76378-458
- 76378-436
- Operating temperature range:
- 0…40 °C (32…104 °F)
- 0…40 °C (32…104 °F)
- 0…40 °C (32…104 °F)
- 0…40 °C (32…104 °F)
- 0…40 °C (32…104 °F)
- 0…40 °C (32…104 °F)
- Communication interface:
- RS232
- RS232
- RS232
- RS232
- RS232
- RS232
- Voltage:
- 100 - 240 VAC
- 100 - 240 VAC
- 100 - 240 VAC
- 100 - 240 VAC
- 100 - 240 VAC
- 100 - 240 VAC
Spécifications
Printer