ContactUsLinkComponent
Zero Dilution Inner Liner, 2 ext.Ø.×73L mm, PerkinElmer
Catalog # CA97019-184
Supplier:  PerkinElmer
undefined
Zero Dilution Inner Liner, 2 ext.Ø.×73L mm, PerkinElmer
Catalog # CA97019-184
Supplier:  PerkinElmer
Supplier Number:  N1011446
Specifications
  • Ext.Ø:
    2.0 mm
  • Description:
    Zero dilution inner liner
  • Environmentally Preferable:
  • Cat. no.:
    CA97019-184
Specifications
About this item
To be used in conjunction with outer liner (N1011447).
  • Not compatible with the Clarus 590/690 Capillary Injectors
  • 73 mm length
Inlet liners for split injection have mixing chambers with tortuous flow paths to allow full vaporization of the sample. Deactivating the surface of these liners prevents active compounds from degrading. Packing the liner with wool will trap non-volatile residue and prevent column contamination when analyzing dirty samples.