Worktables, SPEC-LINE Series with Flat Top, Undershelf Style, Advance Tabco
Fournisseur: Advance Tabco
Certificats
À propos de cet article
Top is furnished with a 2×1" square die embossed NO-DRIP countertop edge with a ¹/₂" return on four sides.
- 14 gauge stainless steel type '304' series top
- Galvanized or stainless steel legs and 18 gauge undershelf
- NSF
To reinforce and maintain a level working surface, 24" wide tables are supplied with two hat channels and 30" and 36" wide tables are supplied with three hat channels. Pre-engineered welded angle adapters insure ease of future drawer installation.
Aluminum die cast 'leg-to-shelf' clamp secures shelf to leg eliminating unsightly nuts and bolts. Undershelf is adjustable.
Frequently Bought Together