Worktables, Stainless Steel, Hinged Door Style with Enclosed Base, Advance Tobaco
Flat top option is furnished with a 1⁵/₈" sanitary rolled rim on the front and 1⁵/₈" square bend edges on the two sides and rear.
- Enclosed cabinet base with hinged doors
- 14 gauge Type 304 stainless steel top
- Bolt-on stainless steel leg
- NSF
: Enclosed base modifications and accessories are available; contact your Avantor representative for more information.
Frequently Bought Together