BRAND Transferpette® -8/-12 Multi-Channel Pipettes, Mechanical, Variable Volume, BrandTech
The Transferpette® S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Due to its perfect ergonomics, the Transferpette® S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands.
- True one-hand operation for right- and left-handed users: Easily set the volume, pipette and eject the tip
- Color code on volume adjustment wheel, shaft and volume-change protection
- Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series
- Easy insertion and ejection of tips due to O-rings on multi-channel pipettes
- Universal tip cone
- Fully autoclavable at 121 °C
The volume-change protection is also easy to operate with gloves on and protects your set volume against accidental adjustment. The Transferpette® S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools. A compatibility table shows which tips fit which device, so that you always order the correct pipette tips.
- Catalog No:
- 76383-116
- 76383-114
- 76383-118
- 76383-108
- 76383-106
- 76383-112
- 76383-110
- 76383-100
- 76383-104
- 76383-102
- Environmentally Preferable:
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
- Sustainable Packaging, Low Manufacturing Impact
Frequently Bought Together