Copper Torch Ignitor Tape for Optima™ 2x00/4x00/5x00/7x00 DV, PerkinElmer
CA97013-028
- Description:Copper torch ignitor tape for Optima™
- Cat. no.:CA97013-028
- Supplier No.:N0775297
The copper foil torch ignitor tape is used with Optima™ 2x00 DV, 4x00 DV, 5x00 DV, and 7x00 DV ICP-OES series instruments. The roll of tape includes 120 pieces.
This copper ignitor tape is also included in the scott/cross-flow II sample introduction kit for Optima™ 2x00/4x00/5x00/7x00 DV (101692-222), the cyclonic/concentric sample introduction kit for Optima™ 2x00/4x00/5x00/7x00 DV (101676-468), and the organics kit for Optima™ 4300 V/5300 V/7300 V(10136-316).