Quartz SUPRASIL® Macro Cylindrical Cells with PTFE Stopper, PerkinElmer
Fournisseur: PerkinElmer
Certificats
À propos de cet article
The quartz SUPRASIL® macro cylindrical cells are used with spectrophotometers that have a larger circular light beam.
- Each cell has an Ext. Ø 22 mm and an Int. Ø 19 mm
Packaging: Each package contains one cell and two windows.
Frequently Bought Together