Variable Angle Reflectance Accessory for PerkinElmer Lambda Spectrometers, PerkinElmer
Variable angle reflectance accessory
Catalog # CA97024-530
Supplier: PerkinElmer
Fournisseur #: B0137314
Variable Angle Reflectance Accessory for PerkinElmer Lambda Spectrometers, PerkinElmer
Catalog # CA97024-530
Supplier: PerkinElmer
Jump to:
Product Details & Documents
The variable angle reflectance accessory is for use with LAMBDA 25/35/45/650/750/850/950/1050. The variable angle reflectance accessory is ideal for relative specular reflectance studies which require data from more than one angle of incidence.
- Wavelength range of 175 to 3300 nm and locates in standard sample compartment
- Angular range from 15 to 70° with an accuracy of ±1°
Typical applications include the study of how the reflectivity of a sample changes with change of incident angle.
Spécifications
- Désignation:Variable angle reflectance accessory