VWR® Transfer Caps for Shaker Flasks
These transfer caps are for the VWR® 5 L shaker flasks. Many caps are available for various applications.
- Inverted transfer caps use gravity to transfer which maintains higher culture viability
- Bi-directional caps use a standard pump to transfer culture or media
Transfer caps allow quick and sterile cell transfer between flask and vessel.
Frequently Bought Together