Order Entry
Canada
ContactUsLinkComponent
Toad Skeleton Plastomount
Catalogue # CA470324-630
undefined
Toad Skeleton Plastomount
Catalogue # CA470324-630
Supplier Number:  TE37

Spécifications

  • Width:
    3"
  • Height:
    1"
  • Length:
    3"
  • Cat. no.:
    CA470324-630
  • No. of Parts:
    1

Spécifications

À propos de cet article

Preserved specimens in resin material.

  • High quality display mount

This specimen block has a REAL Toad Skeleton (Bufo bufo gargarizans) encased. The Skeleton is preserved and encased in clear resin material. It can be observed from every side to see details.

Each specimen is unique and different and may differ from the picture shown.

Delivery: This Skeleton comes in a black box and a detailed Information Card.