ContactUsLinkComponent
Phyllotaxis Sorts Plastomount
  CA470324-592
undefined
Phyllotaxis Sorts Plastomount
  CA470324-592
 :  T703
  • Width:
    3"
  • Height:
    1"
  • Length:
    6.5"
  • Cat. no.:
    CA470324-592
Preserved specimens in resin material.
  • High quality display mount
This kit looks at plants veins. There is one specimen block included detailing the 3 types of Plant veins (Morus alba, Phyllostachys aureosulcata f. spectabilis . & GinkgobilobaL) Specimens are preserved and encased in clear resin material. They can be observed from every side to see details.
 : Information sheet is included.