Ruler, 6" Excelta®
Ruler, straight, 2 star quality, Plastic, 152 mm (6")
Catalog # 75880-502
Supplier: Excelta
Supplier #: RU-1
Jump to:
Product Details & Documents
The RU-1 is a 6" straight plastic ruler.
Specifications
- Désignation:Ruler, straight, 2 star quality
- Longueur:152 mm (6")
- Matière:Plastic