Continuity Tester, Excelta Corp®
Continuity tester, 3 star quality, straight, 37.5×75×100 mm (1.5×3×4")
Catalog # 75880-498
Supplier: Excelta
: PB-1
Jump to:
Product Details & Documents
Quick discernment of point-to-point continuity.
- Quick and easy tester for continuity
- Replaceable probe available
Our popular 'PB-1' Series tester affords the operator quick discernment of point-to-point continuity. As a transistor and diode tester, it analyzes 'in circuit' junctions for go-no/go normality. It indicates leakage, shorted or open conditions of transistors.
Spécifications
- Désignation:Continuity tester, 3 star quality, straight
- Dimensions:37.5×75×100 mm (1.5×3×4")
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Tester, straight replacement probe, 3 star qualityTester, straight replacement probe, 3 star quality89411-974Log in pour la disponibilitéEach - $63.15