Forceps, Gemologist, Standard, Excelta Corp®
Marketsource
Supplier:   Excelta
Forceps, Gemologist, Standard, Excelta Corp®
Supplier:  Excelta
Marketsource
Certificates
Forceps, Gemologist, Standard, Excelta Corp®
Forceps, Gemologist, Standard, Excelta Corp®
7 Options Available Below
Product Details & Documents
Designed for handling gem stones.
  • Gemologist forceps for fine gemstones
  • Fluted grips for positive control
  • Stainless steel construction
These straight Gemologist forceps are designed to handle extra fine gems. The handles are serrated for better control. These are made of AISI 440 Stainless Steel except for the Trisista, which are made of AISI 316L Stainless Steel, which is anti-magnetic.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Overall Length
    Tip Width
    Curvature
    Availability
    Price
  • Forceps, gemologist, 3 star quality
    Stainless steel AISI 440
    159.38 mm (6.375")
    0.4 mm (0.016")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $115.64
    $115.64
    Each
       $115.64
    Supplier Number: JS-2
    Spécifications:
    Marketsource
    Marketsource
  • Forceps, gemologist, 3 star quality
    Stainless steel AISI 440
    159.38 mm (6.375")
    0.63 mm (0.025")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $103.14
    $103.14
    Each
       $103.14
    Supplier Number: JS-3
    Spécifications:
    Marketsource
    Marketsource
  • Forceps, gemologist, 3 star quality
    Stainless steel AISI 440
    159.38 mm (6.375")
    0.88 mm (0.035")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $103.14
    $103.14
    Each
       $103.14
    Supplier Number: JS-4
    Spécifications:
    Marketsource
    Marketsource
  • Forceps, gemologist, 3 star quality
    Stainless steel AISI 440
    159.38 mm (6.375")
    1.19 mm (0.0475")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $123.59
    $123.59
    Each
       $123.59
    Supplier Number: JS-5
    Spécifications:
    Marketsource
    Marketsource
  • Forceps, gemologist, 3 star quality
    Stainless steel AISI 440
    159.38 mm (6.375")
    1.78 mm (0.071")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $103.14
    $103.14
    Each
       $103.14
    Supplier Number: JS-7
    Spécifications:
    Marketsource
    Marketsource
  • Forceps, gemologist, 3 star quality, serrated, black matte finish
    Stainless steel 316L, anti-magnetic
    162.50 mm (6.5")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $111.37
    $111.37
    Each
       $111.37
    Supplier Number: TRISISTA3
  • Forceps, gemologist, 3 star quality, serrated
    Stainless steel 316L, anti-magnetic
    162.50 mm (6.5")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $93.14
    $93.14
    Each
       $93.14
    Supplier Number: TRISISTA7
    Spécifications:
    Marketsource
    Marketsource