Forceps, Gemologist, Slide Lock, Excelta Corp® | Avantor
Forceps, Gemologist, Slide Lock, Excelta Corp®
Marketsource
Supplier:   Excelta
Forceps, Gemologist, Slide Lock, Excelta Corp®
Supplier:  Excelta
Marketsource
Certificates
Forceps, Gemologist, Slide Lock, Excelta Corp®
Forceps, Gemologist, Slide Lock, Excelta Corp®
4 Options Available Below
Product Details & Documents
Ideal for handling gem stones - lock holds the gem securely
  • Slide lock allows gem to be held indefinitly
  • Various sizes available
  • Stainless steel corrosion resistant
These gemologist straight medium point tweezers are ideal for handling medium gems. The handles are serrated for better control. They feature a 'slide-lock' to lock the tweezrers in place while holding the gem.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Overall Length
    Finish
    Tip Width
    Curvature
    Availability
    Price
  • Forceps, gemologist, 3 star quality, slide lock
    Stainless steel AISI 440
    159.38 mm (6.375")
    Black oxide
    0.89 mm (0.0355")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $160.45
    $160.45
    Each
       $160.45
    La quantité minimale de commande est de 1
    Supplier Number: JLB-1
    Spécifications:
    Marketsource
    Marketsource
  • Forceps, gemologist, 3 star quality, slide lock
    Stainless steel AISI 440
    159.38 mm (6.375")
    Black oxide
    1.78 mm (0.071")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $160.45
    $160.45
    Each
       $160.45
    La quantité minimale de commande est de 1
    Supplier Number: JLB-3
    Spécifications:
    Marketsource
    Marketsource
  • Forceps, gemologist, 3 star quality, slide lock
    Stainless steel AISI 440
    159.38 mm (6.375")
    Stainless finish
    1.19 mm (0.0475")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $132.72
    $132.72
    Each
       $132.72
    La quantité minimale de commande est de 1
    Supplier Number: JLS-3
    Spécifications:
    Marketsource
    Marketsource
  • Forceps, gemologist, 3 star quality, slide lock
    Stainless steel AISI 440
    159.38 mm (6.375")
    Stainless finish
    1.78 mm (0.071")
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $124.99
    $124.99
    Each
       $124.99
    La quantité minimale de commande est de 1
    Supplier Number: JLS-6
    Spécifications:
    Marketsource
    Marketsource